Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3,5-Difluorophenoxy)-3-bromoaniline 98%

Supplier: APOLLO SCIENTIFIC

PC303421-10G PC303421-1G PC303421-5G
77414-392EA 394.28 USD InStock
77414-392 77414-394 77414-396
4-(3,5-Difluorophenoxy)-3-bromoaniline 98%
4-(3,5-Difluorophenoxy)-3-bromoaniline


Formula: C₁₂H₈BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD17121998
CAS Number: 1483390-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty