Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2,3-difluorophenylacetonitrile 97%

Supplier: APOLLO SCIENTIFIC

PC303433-1G PC303433-5G
77414-398EA 137.54 USD InStock
77414-398 77414-402
6-Bromo-2,3-difluorophenylacetonitrile 97%
6-Bromo-2,3-difluorophenylacetonitrile


Formula: C₈H₄BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD26686146
CAS Number: 1517576-71-4
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty