Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,2,3,3,3-Hexafluoropropyl 1H,1H-heptafluorobutyl ether 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC450184-5G PC450184-1G
77415-474EA 1354.8 USD InStock
77415-474 77415-472
1,1,2,3,3,3-Hexafluoropropyl 1H,1H-heptafluorobutyl ether 95%
1,1,2,3,3,3-Hexafluoropropyl 1H,1H-heptafluorobutyl ether


Formula: C₇H₃F₁₃O
Boiling Pt: 105 °C
Density: 1.6292 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD22123962
CAS Number: 1184-97-0
UN: 3271
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty