Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluorotetrahydro-2H-pyran-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Danger

PC450285-1G PC450285-5G
77415-528EA 104.94 USD InStock
77415-528 77415-530
Perfluorotetrahydro-2H-pyran-2-carboxylic acid
Perfluorotetrahydro-2H-pyran-2-carboxylic acid


Formula: C₆HF₉O₃
Storage Temperature: Refrigerator
MDL Number: MFCD28054326
CAS Number: 2248063-91-2
UN: 1760
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty