Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Difluoroethyl chloroformate

Supplier: APOLLO SCIENTIFIC
Danger

PC450490-5G PC450490-1G PC450490-25G
77415-554EA 345.4 USD InStock
77415-554 77415-550 77415-552
2,2-Difluoroethyl chloroformate
2,2-Difluoroethyl chloroformate


Formula: C₃H₃ClF₂O₂
Boiling Pt: 72…74 °C (150 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD24141311
CAS Number: 111022-16-3
UN: 2742
ADR: 6.1+3+8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty