Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-(1,2-Difluorovinyl)trimethylsilane 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC450572-1G PC450572-250MG
77415-582EA 812 USD InStock
77415-582 77415-584
(Z)-(1,2-Difluorovinyl)trimethylsilane 97%
(Z)-(1,2-Difluorovinyl)trimethylsilane


Formula: C₅H₁₀F₂Si
Storage Temperature: Ambient
MDL Number: MFCD30062987
CAS Number: 17757-16-3
UN: 1993
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty