Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Difluoromethyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC48367-250MG PC48367-1G
77415-754EA 90.2 USD InStock
77415-754 77415-752
4-(Difluoromethyl)benzonitrile
4-(Difluoromethyl)benzonitrile


Formula: C₈H₅F₂N
MW: 153.129 g/mol
Boiling Pt: 80…90 °C (20 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD23843788
CAS Number: 55805-10-2
UN: 3276
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty