Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-2-(trifluoromethyl)butanoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4863-1G PC4863-250MG
77415-782EA 79.1 USD InStock
77415-782 77415-784
2-Hydroxy-2-(trifluoromethyl)butanoic acid 97%
2-Hydroxy-2-(trifluoromethyl)butanoic acid


Formula: C₅H₇F₃O₃
Boiling Pt: 195…198 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00190638
CAS Number: 72114-82-0
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty