Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(trifluoromethoxy)phenylacetylene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC48787-1G PC48787-250MG
77415-796EA 109.02 USD InStock
77415-796 77415-798
3-Bromo-5-(trifluoromethoxy)phenylacetylene 98%
3-Bromo-5-(trifluoromethoxy)phenylacetylene


Formula: C₉H₄BrF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD31726319
CAS Number: 2366994-52-5
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty