Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-nitrothiophenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC49621-1G PC49621-250MG
77415-934EA 242.48 USD InStock
77415-934 77415-936
4-Fluoro-2-nitrothiophenol 95%
4-Fluoro-2-nitrothiophenol


Formula: C₆H₄FNO₂S
MW: 173.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22481385
CAS Number: 654063-68-0
UN: 3335
ADR: 9 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty