Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(bromomethyl)-5-fluorophenylacetate

Supplier: APOLLO SCIENTIFIC

PC501140-100MG PC501140-1G PC501140-250MG
77416-040EA 137.54 USD InStock
77416-040 77416-042 77416-044
Methyl 3-(bromomethyl)-5-fluorophenylacetate
Methyl 3-(bromomethyl)-5-fluorophenylacetate


Formula: C₁₀H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28348474
CAS Number: 1980044-80-1
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty