Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-1-(trifluoromethoxy)benzene 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC501509-1G PC501509-250MG PC501509-5G
77416-086EA 131.42 USD InStock
77416-086 77416-088 77416-090
3,5-Difluoro-1-(trifluoromethoxy)benzene 99%
3,5-Difluoro-1-(trifluoromethoxy)benzene


Formula: C₇H₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD22581043
CAS Number: 1404194-50-8
UN: 1993
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty