Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethoxy)phenylacetylene 95+%

Supplier: APOLLO SCIENTIFIC
Warning

PC53008-1G PC53008-250MG PC53008-5G
77416-596EA 69.28 USD InStock
77416-596 77416-598 77416-600
2-(Trifluoromethoxy)phenylacetylene 95+%
2-(Trifluoromethoxy)phenylacetylene


Formula: C₉H₅F₃O
MW: 186.131 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03839994
CAS Number: 886363-40-2
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty