Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanoic acid

Supplier: APOLLO SCIENTIFIC
Danger

PC53110-1G PC53110-250MG
77416-634EA 503.3 USD InStock
77416-634 77416-636
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanoic acid
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanoic acid


Formula: C₈H₃F₁₃O₂
MW: 378.087 g/mol
Melting Pt: 55 °C
Density: 2.235 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06203190
CAS Number: 53826-12-3
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty