Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-1-butanol

Supplier: APOLLO SCIENTIFIC
Warning

PC53179-10G PC53179-1G PC53179-5G
77416-648EA 126.34 USD InStock
77416-648 77416-650 77416-652
4-Fluoro-1-butanol
4-Fluoro-1-butanol


Formula: C₄H₉FO
MW: 92.112 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00039555
CAS Number: 372-93-0
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty