Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S)-2-Trifluoromethylpyrrolidine hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC56831-1G PC56831-5G
77416-878EA 32.36 USD InStock
77416-878 77416-880
(2S)-2-Trifluoromethylpyrrolidine hydrochloride 97%
(2S)-2-Trifluoromethylpyrrolidine hydrochloride


Formula: C₅H₈F₃N·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD22380026
CAS Number: 1410903-57-9
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty