Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,2H,2H-Perfluorohexanesulfonic acid

Supplier: APOLLO SCIENTIFIC
Danger

PC6901-1G PC6901-250MG
77417-594EA 771.24 USD InStock
77417-594 77417-596
1H,1H,2H,2H-Perfluorohexanesulfonic acid
1H,1H,2H,2H-Perfluorohexanesulfonic acid


Formula: C₆H₅F₉O₃S
Storage Temperature: Ambient
MDL Number: MFCD14581696
CAS Number: 757124-72-4
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty