Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (cyclopropylmethyl)trifluoroborate 95%

Supplier: APOLLO SCIENTIFIC

PC905207-1G PC905207-250MG
77418-528EA 237.38 USD InStock
77418-528 77418-530
Potassium (cyclopropylmethyl)trifluoroborate 95%
Potassium (cyclopropylmethyl)trifluoroborate


Formula: C₄H₇BF₃·K
MW: 162.003 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11052655
CAS Number: 1356481-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty