Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Pyrazol-3-yl)phenol 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR15021-1G OR15021-500MG OR15021-5G
77405-240EA 55.02 USD InStock
77405-240 77405-242 77405-244
2-(1H-Pyrazol-3-yl)phenol 96%
2-(1H-Pyrazol-3-yl)phenol


Formula: C₉H₈N₂O
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD03093876
CAS Number: 34810-67-8
EINECS: 252-222-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty