Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluoroisopropyl methyl ether 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC4757-100G PC4757-25G PC4757-5G
77415-702EA 496.16 USD InStock
77415-702 77415-704 77415-706
Hexafluoroisopropyl methyl ether 99%
Hexafluoroisopropyl methyl ether


Formula: C₄H₄F₆O
Boiling Pt: 50 °C
Density: 1.39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00221772
CAS Number: 22052-84-2
UN: 3271
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty