Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorogenic acid 98+%

Supplier: APOLLO SCIENTIFIC

OR1166-25G OR1166-5G OR1166-1G
77404-000EA 146.7 USD InStock
77404-000 77404-002 77403-992
Chlorogenic acid 98+%
Chlorogenic acid


Formula: C₁₆H₁₈O₉
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003862
CAS Number: 327-97-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty