Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium(II)(pi-cinnamyl) chloride

Supplier: APOLLO SCIENTIFIC
Danger

IN2814-1G IN2814-250MG IN2814-5G
77401-516EA 128.38 USD InStock
77401-516 77401-518 77401-520
Palladium(II)(pi-cinnamyl) chloride
Palladium(II)(pi-cinnamyl) chloride


Formula: C₁₈H₁₈Cl₂Pd₂
MW: 518.08 g/mol
Melting Pt: 218…220 °C
MDL Number: MFCD16621470
CAS Number: 12131-44-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty