Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenylboronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC
Warning

OR11007-100G OR11007-25G OR11007-500G
77403-060EA 96.78 USD InStock
77403-060 77403-062 77403-064
Phenylboronic acid pinacol ester
Phenylboronic acid pinacol ester


Formula: C₁₂H₁₇BO₂
Melting Pt: 27…31 °C
Flash Pt: 107 °C
MDL Number: MFCD00966985
CAS Number: 24388-23-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty