Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloronicotinamide 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR1214-5G OR1214-1G OR1214-25G
77404-214EA 23.36 USD InStock
77404-214 77404-204 77404-206
2-Chloronicotinamide 97%
2-Chloronicotinamide


Formula: C₆H₅ClN₂O
Melting Pt: 164…167 °C
MDL Number: MFCD00006237
CAS Number: 10366-35-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty