Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dimethoxy-1H-indole-2-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR13450-1G OR13450-250MG OR13450-5G
77404-648EA 247.58 USD InStock
77404-648 77404-650 77404-652
4,5-Dimethoxy-1H-indole-2-carboxylic acid 95%
4,5-Dimethoxy-1H-indole-2-carboxylic acid


Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
Melting Pt: 222…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02664472
CAS Number: 50536-49-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty