Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromothiophenol 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR2222-25G OR2222-5G
77406-844EA 57.06 USD InStock
77406-844 77406-846
2-Bromothiophenol 97%
2-Bromothiophenol


Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 128
Density: 1.573
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004827
CAS Number: 6320-02-1
UN: 2922
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty