Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-cyclopropylbenzoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR70323-1G OR70323-250MG OR70323-5G
77410-998EA 496.16 USD InStock
77410-998 77411-000 77411-002
4-Bromo-3-cyclopropylbenzoic acid 95%
4-Bromo-3-cyclopropylbenzoic acid


Formula: C₁₀H₉BrO₂
MW: 241.08 g/mol
MDL Number: MFCD32862612
CAS Number: 2384580-34-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty