Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

But-3-yn-1-amine 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR72152-250MG OR72152-5G
77411-080EA 67.26 USD InStock
77411-080 77411-082
But-3-yn-1-amine 96%
But-3-yn-1-amine


Formula: C₄H₇N
Boiling Pt: 100…103 °C
Density: 0.844
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997741
CAS Number: 14044-63-4
UN: 2733
ADR: 3+8,II
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty