Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR918031-10G OR918031-1G OR918031-5G
77411-712EA 363.72 USD InStock
77411-712 77411-714 77411-716
4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde 98%
4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde


Formula: C₆H₇N₃OS
MW: 169.2 g/mol
Melting Pt: 180…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669438
CAS Number: 770-31-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty