Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(lauroyloxy)dioctyltin 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR918257-100G OR918257-1G OR918257-25G
77411-722EA 135.5 USD InStock
77411-722 77411-724 77411-726
Bis(lauroyloxy)dioctyltin 96%
Bis(lauroyloxy)dioctyltin


Formula: C₄₀H₈₀O₄Sn
MDL Number: MFCD00026557
CAS Number: 3648-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty