Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(tert-Butyldimethylsilyloxy)-1-methoxyethene

Supplier: APOLLO SCIENTIFIC
Danger

OR923452-1G OR923452-5G
77411-860EA 55.02 USD InStock
77411-860 77411-862
1-(tert-Butyldimethylsilyloxy)-1-methoxyethene
1-(tert-Butyldimethylsilyloxy)-1-methoxyethene


Formula: C₉H₂₀O₂Si
MW: 188.34 g/mol
Boiling Pt: 62/9mm °C
Density: 0.863
Flash Pt: 52 °C
MDL Number: MFCD02683524
CAS Number: 77086-38-5
UN: 1993
ADR: 3 III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty