Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lead tetraacetate 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR924077-100G OR924077-25G
77411-876EA 85.58 USD InStock
77411-876 77411-878
Lead tetraacetate 96%
Lead tetraacetate


Formula: (H₃CCOO)₄Pb
Melting Pt: 175…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008693
CAS Number: 546-67-8
UN: 1616
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty