Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluoro-2-methoxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Danger

PC104325-1G PC104325-250MG PC104325-5G
77412-900EA 344.36 USD InStock
77412-900 77412-902 77412-904
3,4-Difluoro-2-methoxybenzaldehyde
3,4-Difluoro-2-methoxybenzaldehyde


Formula: C₈H₆F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD04115951
CAS Number: 1023023-24-6
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty