Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluoro-2-methoxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC500670-1G PC500670-250MG PC500670-5G
77416-024EA 552.2 USD InStock
77416-024 77416-026 77416-028
4-Bromo-3-fluoro-2-methoxybenzaldehyde
4-Bromo-3-fluoro-2-methoxybenzaldehyde


Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28098074
CAS Number: 1695567-81-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty