Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-nitrobenzenesulphonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC501630-250MG PC501630-5G
77416-118EA 74.38 USD InStock
77416-118 77416-120
4-Fluoro-2-nitrobenzenesulphonyl chloride
4-Fluoro-2-nitrobenzenesulphonyl chloride


Formula: C₆H₃ClFNO₄S
Boiling Pt: 150…155 °C
Melting Pt: 44…47 °C
MDL Number: MFCD03618461
CAS Number: 568586-10-7
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty