Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)-5-vinylpyridine

Supplier: APOLLO SCIENTIFIC
Danger

PC502015-1G PC502015-5G
77416-150EA 816.08 USD InStock
77416-150 77416-152
2-(Trifluoromethyl)-5-vinylpyridine
2-(Trifluoromethyl)-5-vinylpyridine


Formula: C₈H₆F₃N
MDL Number: MFCD19689780
CAS Number: 1133879-76-1
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty