Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluoro-3-hydroxybenzonitrile

Supplier: APOLLO SCIENTIFIC

PC57505-1G PC57505-5G
77417-020EA 496.16 USD InStock
77417-020 77417-022
2-Chloro-6-fluoro-3-hydroxybenzonitrile
2-Chloro-6-fluoro-3-hydroxybenzonitrile


Formula: C₇H₃ClFNO
MW: 171.56 g/mol
MDL Number: MFCD26687881
CAS Number: 1232774-20-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty