Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromo-2-chloro-6-fluorophenyl)methanol

Supplier: APOLLO SCIENTIFIC
Warning

PC57828-1G PC57828-5G
77417-074EA 578.68 USD InStock
77417-074 77417-076
(4-Bromo-2-chloro-6-fluorophenyl)methanol
(4-Bromo-2-chloro-6-fluorophenyl)methanol


Formula: C₇H₅BrClFO
MW: 239.469 g/mol
MDL Number: MFCD25460197
CAS Number: 1823343-34-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty