Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloro-6-fluorobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC57829-1G PC57829-25G PC57829-5G
77417-078EA 578.68 USD InStock
77417-078 77417-080 77417-082
4-Bromo-2-chloro-6-fluorobenzyl bromide
4-Bromo-2-chloro-6-fluorobenzyl bromide


Formula: C₇H₄Br₂ClF
MW: 302.37 g/mol
MDL Number: MFCD25460199
CAS Number: 1823959-60-9
UN: 3261
ADR: 8 III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty