Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodoperfluorbutane tech

Supplier: APOLLO SCIENTIFIC
Warning

PC99446-100G PC99446-25G PC99446-500G
77418-808EA 57.78 USD InStock
77418-808 77418-810 77418-812
1-Iodoperfluorbutane tech
1-Iodoperfluorbutane


Formula: C₄F₉I
Boiling Pt: 64…66 °C
Melting Pt: –88 °C
Density: 2.01
Flash Pt: > 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001062
CAS Number: 423-39-2
UN: 3082
ADR: 9+EHS,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty