Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(trifluoromethyl)nicotinic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC99459-1G PC99459-250MG PC99459-5G
77418-814EA 1032.06 USD InStock
77418-814 77418-816 77418-818
2-Fluoro-5-(trifluoromethyl)nicotinic acid
2-Fluoro-5-(trifluoromethyl)nicotinic acid


Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
MDL Number: MFCD16611307
CAS Number: 1227512-36-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty