Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4,6-difluorobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC99504-1G PC99504-25G PC99504-5G
77418-842EA 578.68 USD InStock
77418-842 77418-844 77418-846
2-Chloro-4,6-difluorobenzyl bromide
2-Chloro-4,6-difluorobenzyl bromide


Formula: C₇H₄BrClF₂
MDL Number: MFCD28750504
CAS Number: 1805526-63-9
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty