Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-chloro-1-fluoro-4-methoxybenzene

Supplier: APOLLO SCIENTIFIC
Warning

PC99570-1G PC99570-5G
77418-866EA 367.8 USD InStock
77418-866 77418-868
2-Bromo-3-chloro-1-fluoro-4-methoxybenzene
2-Bromo-3-chloro-1-fluoro-4-methoxybenzene


Formula: C₇H₅BrClFO
MDL Number: MFCD29043796
CAS Number: 1785613-56-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty