Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluoro-3-iodobenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC99645-1G PC99645-250MG PC99645-5G
77418-900EA 578.68 USD InStock
77418-900 77418-902 77418-904
5-Bromo-2-fluoro-3-iodobenzonitrile
5-Bromo-2-fluoro-3-iodobenzonitrile


Formula: C₇H₂BrFIN
MDL Number: MFCD30829168
CAS Number: 1131632-57-9
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty