Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butylbenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR10947-100G OR10947-25G
77402-986EA 47.88 USD InStock
77402-986 77402-988
tert-Butylbenzene 99%
tert-Butylbenzene


Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 169
Melting Pt: -58
Density: 0.867 g/cm³ (25 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008816
CAS Number: 98-06-6
EINECS: 202-632-4
UN: 2709
ADR: 3 III
Merck Index: 13,01550
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty