Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[3,5-Bis(methylsulfonyl)phenyl]methanol 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR15380-1G OR15380-200MG OR15380-5G
77405-384EA 296.48 USD InStock
77405-384 77405-386 77405-388
[3,5-Bis(methylsulfonyl)phenyl]methanol 95%
[3,5-Bis(methylsulfonyl)phenyl]methanol


Formula: C₉H₁₂O₅S₂
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD04037983
CAS Number: 849924-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty