Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-2-hydroxybenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC1137-250MG PC1137-100MG PC1137-1G
77413-054EA 80.48 USD InStock
77413-054 77413-050 77413-052
3,5-Difluoro-2-hydroxybenzonitrile
3,5-Difluoro-2-hydroxybenzonitrile


Formula: C₇H₃F₂NO
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD08275288
CAS Number: 862088-17-3
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty