Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-4-(trifluoromethyl)benzyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC5001-25G PC5001-5G
77415-994EA 825.24 USD InStock
77415-994 77415-996
2-Nitro-4-(trifluoromethyl)benzyl chloride 98%
2-Nitro-4-(trifluoromethyl)benzyl chloride


Formula: C₈H₅ClF₃NO₂
MW: 239.58 g/mol
Melting Pt: 42…45 °C
Storage Temperature: Ambient
MDL Number: MFCD01631694
CAS Number: 225656-59-7
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty