Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-methoxybenzenesulfonic acid ≥95.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

A0491-500G A0491-25G
TCA0491-500GEA 235 USD InStock
TCA0491-500G TCA0491-025G
3-Amino-4-methoxybenzenesulfonic acid ≥95.0% (by titrimetric analysis)
3-Amino-4-methoxybenzenesulphonic acid
CAS Number: 98-42-0
MDL Number: MFCD00035759
Molecular Formula: C7H9NO4S
Molecular Weight: 203.21
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 305

Other Product Names / Synonyms: 2-Aminoanisole-4-sulfonic Acid, 2-Methoxyaniline-5-sulfonic Acid


Formula: C₇H₉NO₄S
Melting Pt: 305 °C
MDL Number: MFCD00035759
CAS Number: 98-42-0
UN: 2585
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=671e57889cf504c8ec04b98d6f99be547addbf1e&vid=671e57889cf504c8ec04b98d6f99be547addbf1e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty