0210142310
0210142325
0210142380
IC10142310EA
36.36
USD
InStock
IC10142310
IC10142325
IC10142380
Creatinine, anhydrous 98.5-102%, white powder
Creatinine
Creatinine, the end product of creatine catabolism is a normal constituent of urine; daily output about 25 mg/kg body weight. The increased amounts in the urine are typically associated with substantially impaired renal function. Also found together with creatine in muscle tissues and blood. It reacts with picric acid under alkaline conditions to form a Janovski complex. The rate of formation of the colored complex, measured at 480-520 nm is proportional to the creatinine concentration.
Creatinine is a useful marker for normalizing levels of other molecules found in urine and the altered creatinine levels may be associated with conditions that result in decreased renal blood flow, such as diabetes and cardiovascular disease.
- Anhydrous Grade
- Presentation: White Powder
- Soluble in 12 parts water; slightly soluble in ethanol; practically insoluble in acetone, ether, chloroform.
- Storage temperature: Room Temperature
Creatinine, the end product of creatine catabolism is a normal constituent of urine; daily output about 25 mg/kg body weight. The increased amounts in the urine are typically associated with substantially impaired renal function. Also found together with creatine in muscle tissues and blood. It reacts with picric acid under alkaline conditions to form a Janovski complex. The rate of formation of the colored complex, measured at 480-520 nm is proportional to the creatinine concentration.
Creatinine is a useful marker for normalizing levels of other molecules found in urine and the altered creatinine levels may be associated with conditions that result in decreased renal blood flow, such as diabetes and cardiovascular disease.